CAS RN 2765-11-9
IUPAC Name:
pentadecanal
Synonyms:
pentadecanal
Molecular Weight (g/mol):
226.4
Molecular Formula:
C15H30O
InChi Key:
XGQJZNCFDLXSIJ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCC=O
1
–
1
of
1
results
1
Pentadecanal 97.0+%, TCI America™
CAS: 2765-11-9 Molecular Formula: C15H30O Molecular Weight (g/mol): 226.40 InChI Key: XGQJZNCFDLXSIJ-UHFFFAOYSA-N Synonym: Pentadecanaldehyde, Pentadecyl Aldehyde PubChem CID: 17697 ChEBI: CHEBI:17302 IUPAC Name: pentadecanal SMILES: CCCCCCCCCCCCCCC=O