accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2766-17-8

IUPAC Name: ethyl 2-(2-{[(benzyloxy)carbonyl]amino}-3-methylbutanamido)acetate
Synonyms: ethyl 2-(2-{[(benzyloxy)carbonyl]amino}-3-methylbutanamido)acetate
Molecular Weight (g/mol): 336.39
Molecular Formula: C17H24N2O5
InChi Key: FZTGKCOLBDXREB-UHFFFAOYNA-N
SMILES: CCOC(=O)CNC(=O)C(NC(=O)OCC1=CC=CC=C1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)