accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrHSnHH3CCH3H3C

CAS RN 2767-54-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 287.82
Molecular Formula: C6H17BrSn
InChi Key: MXGZXTQIURJKMF-UHFFFAOYSA-N
SMILES: Br.CC[SnH](CC)CC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)