accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2767-91-1

IUPAC Name: 4-(pyridin-4-yl)morpholine
Synonyms: 4-(pyridin-4-yl)morpholine
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: QJWQYVJVCXMTJP-UHFFFAOYSA-N
SMILES: C1CN(CCO1)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)