accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2768-42-5

IUPAC Name: (3R)-3-hydroxy-3-phenylpropanoate
Synonyms: (3R)-3-hydroxy-3-phenylpropanoate
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H9O3
InChi Key: AYOLELPCNDVZKZ-MRVPVSSYSA-M
SMILES: O[C@H](CC([O-])=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results