accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNC

CAS RN 2769-64-4

IUPAC Name: 1-isocyanobutane
Synonyms: N-butyl isocyanide n-butyl isocyanide n-butylisocyanide butyl isocyanide
Molecular Weight (g/mol): 83.13
Molecular Formula: C5H9N
InChi Key: FSBLVBBRXSCOKU-UHFFFAOYSA-N
SMILES: CCCC[N+]#[C-]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)