accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2769-71-3

IUPAC Name: 2-isocyano-1,3-dimethylbenzene
Synonyms: 2-isocyano-1,3-dimethylbenzene
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: DNJLFZHMJDSJFN-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1[N+]#[C-]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)