accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27692-74-6

IUPAC Name: 4-[(pyridin-4-yl)methyl]aniline
Synonyms: 4-(pyridin-4-ylmethyl)aniline
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: WZXYYQHVDJMIFF-UHFFFAOYSA-N
SMILES: NC1=CC=C(CC2=CC=NC=C2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results