accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHNH3CCH3

CAS RN 277299-70-4

IUPAC Name: 2-(3,4-dimethylphenyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one
Synonyms: 2-(3,4-dimethylphenyl)-5-methyl-1H-pyrazol-3-one
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: HBWBJCSXUJIDGN-UHFFFAOYSA-N
SMILES: CC1=CC(=O)N(N1)C1=CC(C)=C(C)C=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)