accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(S)(R)OH(S)O(S)(S)HOHOOOOOOHHOOH

CAS RN 27740-01-8

IUPAC Name: (2S,3S,4S,5R,6S)-6-{[5,6-dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl]oxy}-3,4,5-trihydroxyoxane-2-carboxylic acid
Synonyms: scutellarin scutellarein-7-glucuronide scutellarein-7β-D-glucuronide
Molecular Weight (g/mol): 462.36
Molecular Formula: C21H18O12
InChi Key: DJSISFGPUUYILV-ZFORQUDYSA-N
SMILES: O[C@@H]1[C@@H](O)[C@H](OC2=C(O)C(O)=C3C(=O)C=C(OC3=C2)C2=CC=C(O)C=C2)O[C@@H]([C@H]1O)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)