accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27757-85-3

IUPAC Name: (thiophen-2-yl)methanaminium
Synonyms: thiophen-2-ylmethanaminium
Molecular Weight (g/mol): 114.19
Molecular Formula: C5H8NS
InChi Key: FKKJJPMGAWGYPN-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=CS1
Boiling Point 
  • (3)
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)