accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27772-62-9

IUPAC Name: ethyl 2-methyl-3-oxopropanoate
Synonyms: ethyl 2-methyl-3-oxopropanoate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: VVCYNVCCODBCOE-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results