accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 277756-46-4

IUPAC Name: 1-(trifluoromethyl)cyclopropane-1-carboxylic acid
Synonyms: 1-(trifluoromethyl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 154.09
Molecular Formula: C5H5F3O2
InChi Key: SKCBKBCACWDALV-UHFFFAOYSA-N
SMILES: OC(=O)C1(CC1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)