accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27776-21-2

IUPAC Name: dihydrogen 2-(1-{2-[2-(4,5-dihydro-1H-imidazol-2-yl)propyl]diazen-1-yl}propan-2-yl)-4,5-dihydro-1H-imidazole dichloride
Synonyms: dihydrogen 2-(1-{2-[2-(4,5-dihydro-1H-imidazol-2-yl)propyl]diazen-1-yl}propan-2-yl)-4,5-dihydro-1H-imidazole dichloride
Molecular Weight (g/mol): 323.27
Molecular Formula: C12H24Cl2N6
InChi Key: ZVVWZIADMMTPBX-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC(CN=NCC(C)C1=NCCN1)C1=NCCN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results