accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2778-42-9

IUPAC Name: 1,3-bis(2-isocyanatopropan-2-yl)benzene
Synonyms: 1,3-bis(2-isocyanatopropan-2-yl)benzene
Molecular Weight (g/mol): 244.29
Molecular Formula: C14H16N2O2
InChi Key: AZYRZNIYJDKRHO-UHFFFAOYSA-N
SMILES: CC(C)(N=C=O)C1=CC(=CC=C1)C(C)(C)N=C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results