accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOON

CAS RN 27810-64-6

IUPAC Name: isoquinoline-5-carboxylic acid
Synonyms: isoquinoline-5-carboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: ZIPLFLRGHZAXSJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CN=CC2=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)