accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2783-17-7

IUPAC Name: dodecane-1,12-diamine
Synonyms: 1,12-dodecamethylenediamine
Molecular Weight (g/mol): 200.37
Molecular Formula: C12H28N2
InChi Key: QFTYSVGGYOXFRQ-UHFFFAOYSA-N
SMILES: NCCCCCCCCCCCCN
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)