CAS RN 27833-64-3
IUPAC Name:
13-chloro-10-(4-methylpiperazin-1-yl)-2-oxa-9-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene; butanedioic acid
Synonyms:
loxapine succinate
Molecular Weight (g/mol):
445.9
Molecular Formula:
C22H24ClN3O5
InChi Key:
YQZBAXDVDZTKEQ-UHFFFAOYSA-N
SMILES:
OC(=O)CCC(O)=O.CN1CCN(CC1)C1=NC2=CC=CC=C2OC2=CC=C(Cl)C=C12
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Loxapine succinate
Thermo Scientific Chemicals Loxapine succinate
CAS: 27833-64-3 Molecular Formula: C18H18ClN3O·C4H6O4