accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFOHFFFFF

CAS RN 2787-79-3

IUPAC Name: 2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenol
Synonyms: 2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenol
Molecular Weight (g/mol): 234.07
Molecular Formula: C7HF7O
InChi Key: HZQGKHUTYHEFBT-UHFFFAOYSA-N
SMILES: OC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)