accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27876-94-4

IUPAC Name: (2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid
Synonyms: crocetin natural yellow 6 crocetin sodium salt 8,8'-diapocarotenedioic acid
Molecular Weight (g/mol): 328.41
Molecular Formula: C20H24O4
InChi Key: PANKHBYNKQNAHN-MQQNZMFNSA-N
SMILES: C\C(\C=C\C=C(/C)C(O)=O)=C/C=C/C=C(\C)/C=C/C=C(\C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results