accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2788-86-5

IUPAC Name: 2-(4-chlorophenyl)oxirane
Synonyms: 2-(4-chlorophenyl)oxirane
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: IBWLXNDOMYKTAD-UHFFFAOYNA-N
SMILES: ClC1=CC=C(C=C1)C1CO1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)