CAS RN 278803-20-6
IUPAC Name:
5-(pyridin-3-yl)thiophene-2-carboxylic acid
Synonyms:
5-(pyridin-3-yl)thiophene-2-carboxylic acid
Molecular Weight (g/mol):
205.23
Molecular Formula:
C10H7NO2S
InChi Key:
HENDYXUZFUWIMG-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(S1)C1=CN=CC=C1