accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2NOOCl

CAS RN 2789-25-5

IUPAC Name: 2-chloro-3-nitropyridin-4-amine
Synonyms: 2-chloro-3-nitropyridin-4-amine
Molecular Weight (g/mol): 173.56
Molecular Formula: C5H4ClN3O2
InChi Key: PDQAWJXOYURKPI-UHFFFAOYSA-N
SMILES: NC1=C(C(Cl)=NC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results