accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBr

CAS RN 2789-89-1

IUPAC Name: 1-bromo-4-[2-(4-bromophenyl)ethynyl]benzene
Synonyms: 1-bromo-4-[2-(4-bromophenyl)ethynyl]benzene
Molecular Weight (g/mol): 336.03
Molecular Formula: C14H8Br2
InChi Key: FJQGIJIHOXZMMJ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C#CC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)