accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27893-41-0

IUPAC Name: 4-(heptyloxy)benzaldehyde
Synonyms: 4-(heptyloxy)benzaldehyde
Molecular Weight (g/mol): 220.31
Molecular Formula: C14H20O2
InChi Key: YBCKMIZXHKVONZ-UHFFFAOYSA-N
SMILES: CCCCCCCOC1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)