accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27913-99-1

IUPAC Name: 4-(4-formylphenyl)-1-methylpiperazin-1-ium
Synonyms: 4-(4-formylphenyl)-1-methylpiperazin-1-ium
Molecular Weight (g/mol): 205.28
Molecular Formula: C12H17N2O
InChi Key: PFODEVGLOVUVHS-UHFFFAOYSA-O
SMILES: C[NH+]1CCN(CC1)C1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results