CAS RN 2792-42-9
IUPAC Name:
N-[(2E)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene]hydroxylamine
Synonyms:
N-[(2E)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene]hydroxylamine
Molecular Weight (g/mol):
167.25
Molecular Formula:
C10H17NO
InChi Key:
OVFDEGGJFJECAT-DHZHZOJONA-N
SMILES:
CC1(C)C2CCC1(C)\C(C2)=N\O
1
–
1
of
1
results
1
(1R)-Camphor Oxime 98.0+%, TCI America™
CAS: 2792-42-9 Molecular Formula: C10H17NO Molecular Weight (g/mol): 167.25 MDL Number: MFCD00046298,MFCD00210161 InChI Key: OVFDEGGJFJECAT-DHZHZOJONA-N PubChem CID: 124925274 IUPAC Name: N-[(2E)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ylidene]hydroxylamine SMILES: CC1(C)C2CCC1(C)\C(C2)=N\O