accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 279246-65-0

IUPAC Name: 4-(prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane)
Synonyms: 4-(prop-1-en-1-yl)-4'-propyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 248.45
Molecular Formula: C18H32
InChi Key: IASLDLGGHXYCEO-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1CCC(CC1)C=CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results