accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 279261-81-3

CH3OClBOHHO

CAS RN 279261-81-3

IUPAC Name: (3-chloro-4-ethoxyphenyl)boronic acid
Synonyms: 3-chloro-4-ethoxyphenylboronic acid
Molecular Weight (g/mol): 200.43
Molecular Formula: C8H10BClO3
InChi Key: QZMQZKMQXURQNC-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1Cl)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results