accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27973-29-1

IUPAC Name: 1,6-dibromopyrene
Synonyms: 1,6-dibromopyrene
Molecular Weight (g/mol): 360.05
Molecular Formula: C16H8Br2
InChi Key: JRCJYPMNBNNCFE-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CC3=C(Br)C=CC4=CC=C1C2=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results