accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)HOH2N

CAS RN 2799-16-8

IUPAC Name: (2R)-1-aminopropan-2-ol
Synonyms: (R)-1-amino-2-propanol (2R)-(-)-hydroxypropylamine
Molecular Weight (g/mol): 75.11
Molecular Formula: C3H9NO
InChi Key: HXKKHQJGJAFBHI-GSVOUGTGSA-N
SMILES: C[C@@H](O)CN
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (6)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)