accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBrOSOOHOBr

CAS RN 2800-80-8

IUPAC Name: 3,3-bis(3-bromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: 3,3-bis(3-bromo-4-hydroxyphenyl)-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Weight (g/mol): 512.17
Molecular Formula: C19H12Br2O5S
InChi Key: OYCLSQDXZMROJK-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Br)=C1
Quantity 
  • (1)
  • (2)
  • (1)
Color 
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)