accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2800-96-6

IUPAC Name: tin(4+) tetraacetate
Synonyms: tin(4+) tetraacetate
Molecular Weight (g/mol): 354.89
Molecular Formula: C8H12O8Sn
InChi Key: YJGJRYWNNHUESM-UHFFFAOYSA-J
SMILES: [Sn+4].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)