accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28026-96-2

CH3OHHOOO

CAS RN 28026-96-2

IUPAC Name: 3,5-dihydroxy-4-methylbenzoate
Synonyms: 3,5-dihydroxy-4-methylbenzoate
Molecular Weight (g/mol): 167.14
Molecular Formula: C8H7O4
InChi Key: KMRRXSZDSGYLCD-UHFFFAOYSA-M
SMILES: CC1=C(O)C=C(C=C1O)C([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)