accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOONNNSOOON

CAS RN 28048-33-1

IUPAC Name: disodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate
Synonyms: disodium ferrozine(2-)
Molecular Weight (g/mol): 514.44
Molecular Formula: C20H12N4Na2O6S2
InChi Key: DDIHLFCSEHAOJZ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results