accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28059-64-5

NH2

CAS RN 28059-64-5

IUPAC Name: 2-benzylaniline
Synonyms: 2-benzylaniline
Molecular Weight (g/mol): 183.25
Molecular Formula: C13H13N
InChi Key: DWOBGCPUQNFAFB-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1CC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)