accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOOO

CAS RN 28075-50-5

IUPAC Name: cyclohex-1-en-1-yl trifluoromethanesulfonate
Synonyms: cyclohex-1-en-1-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 230.2
Molecular Formula: C7H9F3O3S
InChi Key: WVSCRRLWRRANJY-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CCCCC1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results