accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNBr

CAS RN 280752-68-3

IUPAC Name: 7-bromo-1-methyl-1H-indole
Synonyms: 7-bromo-1-methylindole
Molecular Weight (g/mol): 210.07
Molecular Formula: C9H8BrN
InChi Key: CALOMHQMSHLUJX-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=CC(Br)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results