accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSOOH

CAS RN 28077-41-0

IUPAC Name: 3-(2-methyl-1,3-thiazol-4-yl)benzoic acid
Synonyms: 3-(2-methyl-1,3-thiazol-4-yl)benzoic acid
Molecular Weight (g/mol): 219.26
Molecular Formula: C11H9NO2S
InChi Key: YDPHSMPSNLAMJE-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC(=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results