accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOI

CAS RN 28080-54-8

IUPAC Name: 2-iodo-5-nitropyridine
Synonyms: 2-iodo-5-nitropyridine
Molecular Weight (g/mol): 250
Molecular Formula: C5H3IN2O2
InChi Key: SJXWHBQWFBHASX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(I)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)