accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHN

CAS RN 2812-46-6

IUPAC Name: tert-butyl pyrrolidine-2-carboxylate
Synonyms: tert-butyl pyrrolidine-2-carboxylate
Molecular Weight (g/mol): 171.24
Molecular Formula: C9H17NO2
InChi Key: XJJBXZIKXFOMLP-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C1CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)