accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOO

CAS RN 2815-95-4

IUPAC Name: 3-(2H-1,3-benzodioxol-5-yl)propanoic acid
Synonyms: 3-(2H-1,3-benzodioxol-5-yl)propanoic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: UIYJGLLTSVRSBM-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results