accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2

CAS RN 28163-64-6

IUPAC Name: 2-phenylpropan-1-amine
Synonyms: Β-methylphenethylamine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: AXORVIZLPOGIRG-UHFFFAOYNA-N
SMILES: CC(CN)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results