CAS RN 28170-13-0
IUPAC Name:
benzenecarbothioic S-acid potassium
Synonyms:
benzenecarbothioic acid potassium
Molecular Weight (g/mol):
177.28
Molecular Formula:
C7H6KOS
InChi Key:
LSBMSRUYNGEWKI-UHFFFAOYSA-N
SMILES:
[K].SC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Potassium thiobenzoate, 95%
Potassium thiobenzoate, 95%
CAS: 28170-13-0 Molecular Formula: C7H6KOS Molecular Weight (g/mol): 177.28 MDL Number: MFCD28399074 InChI Key: LSBMSRUYNGEWKI-UHFFFAOYSA-N Synonym: benzenecarbothioic acid potassium PubChem CID: 87087528 IUPAC Name: benzenecarbothioic S-acid;potassium SMILES: [K].SC(=O)C1=CC=CC=C1