accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28179-33-1

IUPAC Name: 2-bromo-1-(4-phenoxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(4-phenoxyphenyl)ethanone
Molecular Weight (g/mol): 291.14
Molecular Formula: C14H11BrO2
InChi Key: RAXTYMXDSNWNJS-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results