accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)O(S)O(R)OH(S)HO(R)HO

CAS RN 2818-58-8

IUPAC Name: (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
Synonyms: phenylgalactoside
Molecular Weight (g/mol): 256.25
Molecular Formula: C12H16O6
InChi Key: NEZJDVYDSZTRFS-YBXAARCKSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@H]1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)