accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNClN

CAS RN 2818-69-1

IUPAC Name: 6-chloro-2-methyl-1H-1,3-benzodiazole
Synonyms: 5-chloro-2-methyl-3H-1,3-benzodiazole
Molecular Weight (g/mol): 166.61
Molecular Formula: C8H7ClN2
InChi Key: NICFDLORAOTXMD-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(Cl)C=C2N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results