CAS RN 2819-05-8
IUPAC Name:
2-tert-butylpropane-1,3-diol
Synonyms:
2-tert-butylpropane-1,3-diol
Molecular Weight (g/mol):
132.2
Molecular Formula:
C7H16O2
InChi Key:
LQPVVSDKTCYYBZ-UHFFFAOYSA-N
SMILES:
CC(C)(C)C(CO)CO
1
–
1
of
1
results
1
Promotions Available
2-tert-Butylpropane-1,3-diol, 98%, Thermo Scientific Chemicals
2-tert-Butylpropane-1,3-diol, 98%, Thermo Scientific Chemicals
CAS: 2819-05-8 MDL Number: MFCD01075745