accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28210-41-5

IUPAC Name: calcium 2-ethenylbenzene-1-sulfonate
Synonyms: kayexalate sodium polystyrene sulfonic acid SPS kionex
Molecular Weight (g/mol): 223.28
Molecular Formula: C8H7CaO3S
InChi Key: IJBYVKWTVVYTPL-UHFFFAOYSA-M
SMILES: [Ca++].[O-]S(=O)(=O)C1=CC=CC=C1C=C
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results