accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3COSCl

CAS RN 28249-77-6

IUPAC Name: N,N-diethyl{[(4-chlorophenyl)methyl]sulfanyl}formamide
Synonyms: saturno epa pesticide chemical code 108401 bolero thiobencarbe C12H16ClNOS benthiocarb saturn siacarb saturn (herbicide) thiobencarb
Molecular Weight (g/mol): 257.78
Molecular Formula: C12H16ClNOS
InChi Key: QHTQREMOGMZHJV-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)SCC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results